Anti-ITGB1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TS2/16]

Supplier: Biogems
11311-50-25
75842-160EA 191.93 USD
75842-160
Anti-ITGB1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TS2/16]
Antibodies
The TS2/16 moncolonal antibody specifically reacts with human CD29 (Integrin beta1), a 130 kDA type I glycoprotein expressed on hematopoietic and non-hematopoietic cells. It forms the VLA-(1-6) molecules with integrin alpha-(1-6) and is involved in the adhesion between cell-cell and cell-matrix. The TS2/16 antibody has been found to activate beta 1 integrins.

Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: TS2/16
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR