Anti-CD19 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1D3]

Supplier: Biogems
11212-80-100
75842-150EA 277.44 USD
75842-150
Anti-CD19 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1D3]
Antibodies
The 1D3 monoclonal antibody specifically reacts with mouse CD19, a 95 kDa transmembrane glycoprotein, a member of the Ig superfamily and a B cell-lineage differentiation antigen expressed by all the B lymphocyte development stages, except for the terminally differentiated plasma cells. CD19 associates with CD21, CD81 and MHC class II to form a multi-molecular complex that initiates the mature B lymphocyte activation by interaction with the B-cell receptors. CD 19 enhances the B cell proliferation, development and activation, and the maturation of memory B cells. In CD19-deficient mice, the generation and maturation of B lymphocytes in the bone marrow and periphery are affected.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 1D3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR