Anti-CD19 Rat Monoclonal Antibody (BG Violet 450) [clone: 1D3]

Supplier: Biogems
11212-40-25
75842-130EA 129.04 USD
75842-130
Anti-CD19 Rat Monoclonal Antibody (BG Violet 450) [clone: 1D3]
Antibodies
The 1D3 monoclonal antibody specifically reacts with mouse CD19, a 95 kDa transmembrane glycoprotein, a member of the Ig superfamily and a B cell-lineage differentiation antigen expressed by all the B lymphocyte development stages, except for the terminally differentiated plasma cells. CD19 associates with CD21, CD81 and MHC class II to form a multi-molecular complex that initiates the mature B lymphocyte activation by interaction with the B-cell receptors. CD 19 enhances the B cell proliferation, development and activation, and the maturation of memory B cells. In CD19-deficient mice, the generation and maturation of B lymphocytes in the bone marrow and periphery are affected. BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 1D3
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR