Anti-CD8a Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: OKT8]

Supplier: Biogems
10121-70-25
75842-094EA 148.36 USD
75842-094
Anti-CD8a Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: OKT8]
Antibodies
The OKT8 monoclonal antibody specifically reacts with human CD8α molecule, a tyPEI transmembrane glycoprotein of 32-34 kDa. CD8α is a member of the Ig superfamily, expressed as a homodimer (CD8αα) or as a heterodimer (CD8αβ). CD8+ αβ T lymphocytes express both CD8αα and CD8αβ, while some T lymphocytes and the natural killer cells express only the homodimers. CD8 binds to MHC class I and influences the development and the activation of T lymphocytes.OKT8, RPA-T8, and HIT8a antibodies do not compete with each other for binding to peripheral leukocytes, meaning that that they do not recognize the same epitoPEor block each other by steric hindrance.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: OKT8
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR