Anti-CD8a Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: Hit8a]

Supplier: Biogems
10111-80-25
75842-046EA 135.51 USD
75842-046
Anti-CD8a Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: Hit8a]
Antibodies
The Hit8a monoclonal antibody reacts with the human CD8a molecule, a 32 kDa cell surface receptor expressed either as a heterodimer (CD8 α/β) or as a homodimer (CD8 α/α) on the majority of thymocytes, a subpopulation of mature T cells, and natural killer cells. CD8 interacts with the major histocompatibility complex class I (MHC class I) molecules on antigen-presenting cells or epithelial cells. The Hit8a antibody reacts with 13-48% of peripheral lymphocytes, 80% of thymocytes, and a subset of natural killer cells. HIT8a, RPA-T8, and OKT8 antibodies do not compete with each other for binding to peripheral leukocytes, meaning that that they do not recognize the same epitope or block each other by steric hindrance.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: Hit8a
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR