Anti-B3GAT1 Mouse Monoclonal Antibody [clone: TBO1]

Supplier: BIOGEMS INTERNATIONAL INC.
09611-25-500
75842-024EA 722.75 USD
75842-024
Anti-B3GAT1 Mouse Monoclonal Antibody [clone: TBO1]
Antibodies
The TBO1 monoclonal antibody specifically binds to human CD57, a 110 kDA glycoprotein expressed on a subset of natural killer lymphocytes, cells, neural cells and striated muscle. It is reported that the molecule is involved in cell-matrix interactions and is upregulated in some disease states.

Type: Primary
Antigen: B3GAT1
Clonality: Monoclonal
Clone: TBO1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR