Anti-CD27 Armenian Hamster Monoclonal Antibody [clone: LG.7F9]

Supplier: BIOGEMS INTERNATIONAL INC.
09331-25-500
75842-018EA 672.21 USD
75842-018
Anti-CD27 Armenian Hamster Monoclonal Antibody [clone: LG.7F9]
Antibodies
The LG.7F9 monoclonal antibody binds to CD27, 45kDA molecule in the Tumor Necrosis Factor Receptor (TNFR) superfamily. It is expressed on memory B cells, NK cells, mature T cells, and a subset of thymocytes. Cd27 is involved in the interactions between B and T cells through its ligand CD70. The LG.7F9 antibody is cross-reactive human, mouse, and rat CD27.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: LG.7F9
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR