Anti-CD27 Armenian Hamster Monoclonal Antibody [clone: LG.3A10]

Supplier: BIOGEMS INTERNATIONAL INC.
09321-25-50
75842-014EA 233.73 USD
75842-014
Anti-CD27 Armenian Hamster Monoclonal Antibody [clone: LG.3A10]
Antibodies
The LG.3A10 monoclonal antibody binds to CD27, 45kDA molecule in the Tumor Necrosis Factor Receptor (TNFR) superfamily. It is expressed on memory B cells, NK cells, mature T cells, and a subset of thymocytes. Cd27 is involved in the interactions between B and T cells through its ligand CD70. The LG.3A10 antibody is cross-reactive with human, mouse and rat CD27.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: LG.3A10
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR