Anti-CD27 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: O323]

Supplier: Biogems
09311-80-100
75842-006EA 309.69 USD
75842-006
Anti-CD27 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: O323]
Antibodies
The O323 monoclonal antibody specifically binds to the human CD27, which is a lymphocyte-specific member of the TNF/NGF-R family, expressed on a subset of human thymocytes and the majority of the mature T cells. CD27 is upregulated by T lymphocytes stimulation, and binds to CD70, enhancing the interaction between T and B lymphocytes. CD27 is a memory marker of B cells.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: O323
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR