Anti-CD27 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: O323]

Supplier: Biogems
09311-77-25
75842-004EA 154.85 USD
75842-004
Anti-CD27 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: O323]
Antibodies
The O323 monoclonal antibody specifically binds to the human CD27, which is a lymphocyte-specific member of the TNF/NGF-R family, expressed on a subset of human thymocytes and the majority of the mature T cells. CD27 is upregulated by T lymphocytes stimulation, and binds to CD70, enhancing the interaction between T and B lymphocytes. CD27 is a marker of memory B cells.

Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: O323
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR