Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 30-F11]

Supplier: Biogems
07512-77-100
75841-978EA 206.45 USD
75841-978
Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 30-F11]
Antibodies
The 30-F11 monoclonal antibody specifically reacts with all isoforms of CD45 and also with the alloantigens CD45.1 and CD45.2 (LCA). CD45 is a transmembrane glycoprotein, expressed by all the hematopoietic cells, except for platelets and mature erythrocytes, which distinguishes the leukocytes from the non-hematopoietic cells. The CD45 molecule is a member of the Protein Tyrosine Phosphatase (PTP) family, because its intracellular region contains two PTP domains. The extracellular region’s variability is caused by different levels of glycosylation, and the splicing of the 4, 5, and 6 exons.The isoforms found in the mouse strains depend on the activation state, maturation stage and cell type, and are very important in B and T lymphocytes antigen receptor signal transduction.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 30-F11
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR