Anti-IL2RA Rat Monoclonal Antibody (BG Violet 450) [clone: PC61.5]

Supplier: Biogems
07312-40-100
75841-938EA 232.27 USD
75841-938
Anti-IL2RA Rat Monoclonal Antibody (BG Violet 450) [clone: PC61.5]
Antibodies
The PC61.5 antibody specifically reacts with mouse CD25, the 55 kDa low-affinity Interleukin-2 Receptor α chain (IL-2R α), expressed on early progenitors of T and B lineage, and on B and T cells. Together with CD122 (IL-2 Receptor β) and CD 132 (IL-2 Receptor γc, the common gamma chain), CD25 forms high-affinity receptor complexes for IL-2. Resting B and T cells and natural killer cells do not express IL-2Rα. Cd25 is also expressed on the dendritic cells, and it enhances lymphocyte differentiation and activation.The PC61.5 antibody blocks the binding of IL-2 to both high-affinity and low-affinity receptors.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: PC61.5
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG1, lambda
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR