CD25 MAB 100tests
Supplier: Biogems
CD25, Monoclonal Antibody, Clone: BC96, Host: Mouse, Species Reactivity: Human, Isotype: IgG1, kappa, Conjugate: APC, Formulation: Phosphate-buffered aqueous solution, 0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2, Application: FC, Size: 100tests