Anti-CD15 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HI98]

Supplier: BIOGEMS INTERNATIONAL INC.
07211-50-25
75841-904EA 160.35 USD
75841-904
Anti-CD15 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HI98]
Antibodies
The HI98 monoclonal antibody specifically reacts with CD15, a 220 kDa carbohydrate moiety also known as SSEA-1, Lewis 3-fucosyl-N-acetyllactosamine (3-FAL). CD15 is expressed on most granulocytes and monocytes to the exclusion of lymphocytes and basophils. It is involved in cell adhesion, chemotaxis, and phagocytosis.

Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: HI98
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR