Anti-CD45RO Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: UCHL1]

Supplier: Biogems
07141-50-25
75841-886EA 90.32 USD
75841-886
Anti-CD45RO Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: UCHL1]
Antibodies
The UCHL1 monoclonal antibody specifically reacts with human CD45RO, a 180 kDa isoform of the leukocyte common antigen CD45. CD45RO is a transmembrane glycoprotein with tyrosine phosphatase activity and is expressed by majority of thymocytes, monocytes, granulocytes, and activated memory T lymphocytes. The subsets of peripheral T lymphocytes can be discriminated by using the CD45RO and CD45RA expressing cells.

Type: Primary
Antigen: CD45RO
Clonality: Monoclonal
Clone: UCHL1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR