Anti-ICAM1 Mouse Monoclonal Antibody [clone: 15.2]

Supplier: BIOGEMS INTERNATIONAL INC.
06611-20-100
75841-846EA 135.51 USD
75841-846
Anti-ICAM1 Mouse Monoclonal Antibody [clone: 15.2]
Antibodies
The 15.2 antibody reacts with the 85-110 kDa intracellular adhesion molecule-1 (ICAM-1), a member of the Ig superfamily which acts as a ligand for the Lymphocyte Function-Associated Antigen-1 (LFA-1). ICAM-1 is also known CD 54 and is expressed on non-hematopoietic cells of vascular endothelial, thymic epithelial, fibroblasts lineages, and on hematopoietic cells like macrophages, mitogen-stimulated T-lymphoblasts, dendritic cells in tonsils, lymph nodes and Peyer’s patches, and germinal center B cells.Inflammatory mediators (IL-1, TNF, IFN-γ) enhance the production of ICAM-1 on fibroblasts and endothelial cells within few hours. Thus, ICAM-1 seems to be the marker of inflammatory reactions.

Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: 15.2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR