Anti-CD34 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4H11]

Supplier: Biogems
06411-60-25
75841-800EA 274.87 USD
75841-800
Anti-CD34 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4H11]
Antibodies
The 4H11 monoclonal antibody specifically reacts with the Class III epitope of CD34 (mucosialin), a transmembrane phosphoglycoprotein. It is expressed on most hematopoietic progenitors, pluripotent stem cells and tumor cells.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR