Anti-CD24 Rat Monoclonal Antibody [clone: M1/69]

Supplier: BIOGEMS INTERNATIONAL INC.
06312-25-500
75841-776EA 677.96 USD
75841-776
Anti-CD24 Rat Monoclonal Antibody [clone: M1/69]
Antibodies
The M1/69 monoclonal antibody specifically reacts with mouse CD24, variably glycosylated GPI-anchored membrane protein. CD24 is known as the Heat Stable Antigen (HAS or HsAG) and is found on epithelial cells, thymocytes, monocytes, erythrocytes, granulocytes, dendritic cells and lymphocytes to the exclusion of NK cells. It is useful in delineating the stages of lymphocyte development and is involved in the costimulation of CD4+ T cells by B cells. CD62P (P-Selectin) is the reported ligand of CD24.

Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: M1/69
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR