CD24 Mab 25 tests
Supplier: Biogems
CD24 Monoclonal Antibody, clone: SN3, Host: mouse, Species reactivity: human, Isotype: IgG1, kappa, Conjugate: FITC, Formulation: Phosphate-buffered aqueous solution, greater than or equal to 0.09% Sodium azide, carrier protein/stabilizer, ph7.2, Application: FC, Size: 25 tests