Anti-CD14 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Sa2-8]

Supplier: Biogems
06212-50-100
75841-762EA 384.13 USD
75841-762
Anti-CD14 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Sa2-8]
Antibodies
The Sa2-8 monoclonal antibody specifically reacts with mouse CD14, a cell surface anchored glycoprotein that is primarily expressed on macrophages and is secreted in certain conditions. CD14 associates with LPS and LBP to form a receptor complex, which signals specifically in response to bacterial lipopolysaccharide (LPS) binding. The Sa2-8 antibody exhibits a weak antagonistic activate in relation to TNF alpha production and NF-kappa B activation.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: Sa2-8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR