CD3e Mab 100 ug
Supplier: Biogems
CD3e Monoclonal Antibody, clone: 145-2C11, Host: Armenian Hamster, Species reactivity: Mouse, Isotype: IgG, Conjugate: PE, Formulation: Phosphate-buffered aqueous solution, greater than or equal to 0.09% Sodium azide, Application: FC, FA, IHC, IF, IP, Storage: 4 deg C, Size: 100 ug