Anti-CD3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 17A2]

Supplier: Biogems
05112-60-100
75841-678EA 187.06 USD
75841-678
Anti-CD3 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 17A2]
Antibodies
The 17A2 monoclonal antibody specifically reacts with the MouseT lymphocytes receptor (TCR) associated CD3 complex, resulting in cellular activation and proliferation. CD3 is expressed by thymocytes and mature lymphocytes, and contains γ, δ, and ε subunits, involved in the assembly, trafficking, and surface expression of T-cell receptor complex. The interaction between the T lymphocytes and the 17A2 antibody can be blocked by the 145-2C11 anti-CD3e antibody, demonstrating that the 17A2 antibody recognizes the CD3 ε chain.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 17A2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR