CD3 Mab 25 tests
Supplier: Biogems
CD3 Monoclonal Antibody, clone: Hit3a, Host: Mouse, Species reactivity: human, Isotype: IgG2b, kappa, Conjugate: PE, Formulation: Phosphate-buffered aqueous solution, greater than or equal to 0.09% Sodium azide, ph7.2, Application: FC, FA, IHC, IP, Size: 25 tests