Anti-CD62L Mouse Monoclonal Antibody [clone: DREG-56]

Supplier: Biogems
04731-25-500
75841-648EA 837.92 USD
75841-648
Anti-CD62L Mouse Monoclonal Antibody [clone: DREG-56]
Antibodies
The DREG-56 monoclonal antibody specifically reacts with human CD62L, also known as L-selectin and LECAM-1. It mediates the migration of lymphocytes to the site of inflammation and their return to the peripheral lymphoid tissues and to the HEV (high endothelial venules). CD62L is expressed on monocytes, NK cells, neutrophils, granulocytes, and subsets of T and B cells. It is reported that the DREG-56 antibody inhibits the binding of lymphocytes to HEV tissue in frozen peripheral tissue.

Type: Primary
Antigen: CD62L
Clonality: Monoclonal
Clone: DREG-56
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR