Anti-CD62L Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: MEL-14]

Supplier: Biogems
04712-77-100
75841-634EA 251.63 USD
75841-634
Anti-CD62L Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: MEL-14]
Antibodies
The MEL-14 monoclonal antibody specifically reacts with L- selectin (CD62L), a receptor with lectin-like and Epidermal Growth Factor-like domains. The weight of the CD62L molecules depend on their origin: 74 kDa (on lymphocytes) or 95 kDa (on neutrophils).In the Mouseorganism, CD62L can be expressed by most thymocytes, on subsets of B and T lymphocytes, monocytes, eosinophils, and neutrophils. The L-selectin binds sulfated, fucosylated, and glycosylated glycoproteins (MadCAM-1, GLYCAM-1, and CD 34). It mediates the migration of lymphocytes to the site of inflammation and their return to the peripheral lymphoid tissues and to the HEV (high endothelial venules). In vitro, L-selectin inhibits this binding and the lymphocyte extravasation into peripheral lymph nodes.

Type: Primary
Antigen: CD62L
Clonality: Monoclonal
Clone: MEL-14
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR