CD31 (PECAM-1) Mab 100 ug
Supplier: Biogems
CD31 (PECAM-1) Monoclonal Antibody, clone: WM59, Host: Mouse, Species reactivity: human, Isotype: IgG1, kappa, Conjugate: SAFIRE Purified, Formulation: Phosphate-buffered aqueous solution, contain carrier protein, ph7.2, Application: FC, IHC, Storage: 4 deg C, Size: 100 ug