CD11c Mab 25 ug
Supplier: Biogems
CD11c Monoclonal Antibody, clone: N418, Host: Armenian Hamster, Species reactivity: Mouse, Isotype: IgG, Conjugate: APC-Cy7, Formulation: Phosphate-buffered aqueous solution, greater than or equal to 0.09% Sodium azide, ph7.2, Application: FC, FA, IHC, IF, IP, Size: 25 ug