Anti-CD11c Armenian Hamster Monoclonal Antibody (BG Violet 450) [clone: N418]

Supplier: BIOGEMS INTERNATIONAL INC.
03212-40-100
75841-524EA 232.14 USD
75841-524
Anti-CD11c Armenian Hamster Monoclonal Antibody (BG Violet 450) [clone: N418]
Antibodies
The N418 monoclonal antibody specifically reacts with the integrin αx chain of the mouse CD11c, which is expressed on dendritic cells, CD4-/CD8+ intestinal intraepithelial lymphocytes (IEL) and some activated T lymphocytes. Low levels of CD11c were detected on mouse splenic natural killer cells and on the monocyte/macrophage lineage cells.CD11c expression is upregulated on IEL and T lymphocytes after activation. It binds to CD54, fibrinogen and iC3b and influences the leukocyte adhesive interactions. The N418 antibody binds to CD11c on splenic dendritic cells of the mouse in the T-dependent areas. It also contributes to the binding of iC3b.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: N418
Conjugation: BG Violet 450
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR