Anti-CD11c Armenian Hamster Monoclonal Antibody (Biotin) [clone: N418]

Supplier: Biogems
03212-30-500
75841-522EA 277.44 USD
75841-522
Anti-CD11c Armenian Hamster Monoclonal Antibody (Biotin) [clone: N418]
Antibodies
The N418 monoclonal antibody specifically reacts with the integrin αx chain of the mouse CD11c, which is expressed on dendritic cells, CD4-/CD8+ intestinal intraepithelial lymphocytes (IEL) and some activated T lymphocytes. Low levels of CD11c were detected on mouse splenic natural killer cells and on the monocyte/macrophage lineage cells.CD11c expression is upregulated on IEL and T lymphocytes after activation. It binds to CD54, fibrinogen and iC3b and influences the leukocyte adhesive interactions. The N418 antibody binds to CD11c on splenic dendritic cells of the mouse in the T-dependent areas. It also contributes to the binding of iC3b.

Type: Primary
Antigen: CD11c
Clonality: Monoclonal
Clone: N418
Conjugation: Biotin
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR