Anti-CD11b Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ICRF44]

Supplier: BIOGEMS INTERNATIONAL INC.
03211-80-100
75841-512EA 309.69 USD
75841-512
Anti-CD11b Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: ICRF44]
Antibodies
The ICRF44 monoclonal antibody specifically reacts with the 165 kDa human adhesion glycoprotein CD11b, which forms, together with the 95 kDa CD18 (integrin β2) a complex known as Mac-1. CD11b is expressed on the surface of activated lymphocytes, a subset of natural killer cells, granulocytes, and monocytes. It functions as a receptor in cell-cell and cell-matrix interactions and as a receptor for iC3b, ICAM-1, ICAM-2, and ICAM-3 intercellular adhesion molecules.

Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: ICRF44
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR