CD11b Mab 100 tests
Supplier: Biogems
CD11b Monoclonal Antibody, clone: ICRF44, Host: Mouse, Species reactivity: human, Isotype: IgG1, kappa, Conjugate: PE, Formulation: Phosphate-buffered aqueous solution, greater than or equal to 0.09% Sodium azide, ph7.2, Application: FC, FA, IF, IP, WB, Size: 100 tests