Anti-CD1d Rat Monoclonal Antibody [clone: 1B1]

Supplier: Biogems
03112-25-100
75841-494EA 358.06 USD
75841-494
Anti-CD1d Rat Monoclonal Antibody [clone: 1B1]
The 1B1 monoclonal antibody specifically reacts with mouse CD1d, a 48kDA type I membrane glycoprotein with a structural homology to MHC class I molecules. CD1d is expressed by mouse leukocytes, epithelial cells, thymocytes, and dendritic cells and is the only molecules in the CD1 family to have been found in the mouse. The 1B1 antibody detects CD1d in association with the Beta2m molecule.

Type: Primary
Antigen: CD1d
Clonality: Monoclonal
Clone: 1B1
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR