Anti-CD1a Mouse Monoclonal Antibody [clone: HI149]

Supplier: BIOGEMS INTERNATIONAL INC.
03111-20-25
75841-480EA 147.46 USD
75841-480
Anti-CD1a Mouse Monoclonal Antibody [clone: HI149]
Antibodies
The HI149 monoclonal antibody specifically reacts with human CD1a, a 49kDA type I membrane glycoprotein. It is expressed on dendritic cells, Langerhans cells, and cortical thymocytes. CD1a plays a role in antigen presentation and ,like MHC Class I, it non-covalently associates with Beta2-microglobulin.

Type: Primary
Antigen: CD1a
Clonality: Monoclonal
Clone: HI149
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR