Anti-CD90 Rat Monoclonal Antibody [clone: G7]

Supplier: Biogems
03032-25-500
75841-476EA 754.56 USD
75841-476
Anti-CD90 Rat Monoclonal Antibody [clone: G7]
Antibodies
The G7 monoclonal antibody specifically reacts with mouse CD90, also known as thymus cell antigen-1 (Thy-1). CD90 is a 25-35 kDA GPI-anchored protein is the smallest member of the Ig superfamily. CD90 is expressed on thymocytes, neurons, hematopoietic stem cells, and T cells. It plays a role in T cell signal transduction and adhesion. The G7 antibody is reported to stimulate IL-2 secretion and T cell proliferation.

Type: Primary
Antigen: CD90
Clonality: Monoclonal
Clone: G7
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2c, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR