Anti-IL-2 (JES6-5H4) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: JES6-5H4]

Supplier: Biogems
86412-60-100
75841-460EA 232.27 USD
75841-460
Anti-IL-2 (JES6-5H4) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: JES6-5H4]
Antibodies
The JES6-5H4 antibody specifically binds to mouse IL-2, a 17 kDa immunoregulatory cytokine and growth factor for T and B lymphocytes, NK cells, monocytes, and macrophages. IL-2 induces the cytotoxic operations of NK cells, activates B cells, and plays a role in the differentiation of T cells. JES6-5H4 seems to have a neutralizing effect on interleukin-2.

Type: Primary
Antigen: IL-2 (JES6-5H4)
Clonality: Monoclonal
Clone: JES6-5H4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR