Anti-TNFA Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP6-XT22]

Supplier: BIOGEMS INTERNATIONAL INC.
84422-80-100
75841-396EA 637.99 USD
75841-396
Anti-TNFA Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP6-XT22]
Antibodies
The MP6-XT22 monoclonal antibody specifically reacts with mouse tumor necrosis factor alpha (TNF-alpha), a 156 amino pro-inflammatory cytokine. It is secreted by various cells including adipocytes, activated monocytes, macrophages, B cells, T cells and fibroblasts. NF-α is cytotoxic to a wide variety of tumor cells and is an essential factor in mediating the immune response against bacterial infections. TNF-α also plays a role in the induction of septic shock, auto immune diseases, rheumatoid arthritis, inflammation, and diabetes. The MP6-XT22 is reported to neutralize the bioactivity of natural or recombinant TNF-alpha and is useful for intracellular immunofluorescent staining.

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: MP6-XT22
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR