Ly-6G (Gr-1) Mab 500 ug
Supplier: Biogems
Ly-6G (Gr-1) Monoclonal Antibody, clone: RB6-8C5, Host: Rat, Species reactivity: Mouse, Isotype: IgG2b, kappa, Conjugate: Purified, Formulation: Phosphate-buffered aqueous solution, greater than or equal to 0.09% Sodium azide, Application: FC, FA, IHC, IP, Size: 500 ug