Anti-CD7 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 4H9]

Supplier: Biogems
49111-50-100
75841-360EA 473.77 USD
75841-360
Anti-CD7 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 4H9]
Antibodies
The 4H9 monoclonal antibody specifically binds to human CD7, a 40 kDa type I transmembrane glycoprotein known as Leu-9 or gp40. CD7 is found on the surfaces of T cells, NK cells, and acute lymphocytic leukemia cells. It is reported to be involved in modulating cytokine secretion and cellular adhesion.

Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: 4H9
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR