Mouse IgG1, kappa Isotype Control (PE (Phycoerythrin)) [clone: MOPC-21]

Supplier: Biogems
44212-60-100
75841-348EA 174.21 USD
75841-348
Mouse IgG1, kappa Isotype Control (PE (Phycoerythrin)) [clone: MOPC-21]
Antibodies
MOPC-21 is a Mousemyeloma protein from the Ig family. It can be used as an Isotype Control for MouseIgG1 antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR