Mouse IgG2b Isotype Control (APC (Allophycocyanin)) [clone: MPC-11]

Supplier: Biogems
43922-80-100
75841-338EA 167.76 USD
75841-338
Mouse IgG2b Isotype Control (APC (Allophycocyanin)) [clone: MPC-11]
Antibodies
The MPC-11 monoclonal antibody is expressed by a cell line adapted from the Merwin Plasmacytoma-11, and has unknown specificity. The MPC-11 line was obtained by intraperitoneal implantation into a BALB/c Mouseof a Millipore diffusion chamber and is used as an Isotype Control for MouseIgG2b antibodies.

Type: Control
Antigen:
Clonality: Monoclonal
Clone: MPC-11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR