SP Bel-Art H-B DURAC® Bi-Metallic Min./Max. Thermometer, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B61301-0300
75841-284CS 491.27 USD
75841-284
SP Bel-Art H-B DURAC® Bi-Metallic Min./Max. Thermometer, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
Maximum/Minimum thermometers display the current temperature and register the maximum and minimum temperatures attained since the last reset
Maximum/Minimum thermometers are for use in high moisture or wet environments including green houses, sample prep areas and water treatment facilities.