POLYVIEW® Plus HRP-DAB (Anti-Rabbit) Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT159-0150
75841-152EA 693.24 USD
75841-152
POLYVIEW® Plus HRP-DAB (Anti-Rabbit) Kit, Enzo Life Sciences
Stains Microbiology Stains
Complete staining kit with highest sensitivity, lowest background.
  • Highest sensitivity, complete staining kit for both IHC and ISH
  • POLYVIEW® Plus reagents' biotin free nanopolymer detection circumvents endogenous background while providing specific antigen localization
  • HIGHDEF® DAB (Brown) HRP provides high intensity color development to deliver sharp, crisp staining
  • Compatible with both manual and automated platforms
The POLYVIEW® Plus HRP-DAB (Anti-Rabbit) Kit, is intended for both IHC and ISH staining. The kit is a non-biotin one step detection system suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and frozen sections. This kit may also be used with blood smears, cytosmears, and cell preparations.
Ordering information: Contains IHC/ISH Peroxidase Block, Antibody Blocking Buffer, POLYVIEW® Plus HRP (Anti-Rabbit) Reagent (Ready-To-Use), HIGHDEF® DAB Chromogen, HIGHDEF® DAB Substrate Buffer, HIGHDEF® Hematoxylin, Antigen Retrieval Reagent, pH 9 (10X).