Hematoxylin, HIGHDEF® used in nuclear staining

Supplier: Enzo Life Sciences

Synonyms: DcH-48, Hydroxybrazilin, (6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol, NSC 2785, Natural Black I, Hydroxybrasilin, (+)-Hematoxylin

ACC106-0100
75841-140EA 70.85 USD
75841-140
Hematoxylin, HIGHDEF® used in nuclear staining
Hematoxylin

Ready-to-use histologic stain with enhancers.


  • Intended for use in the histologic demonstration of nuclear staining
  • Distinctions between normal and ambnormal nucleus
  • Histologic stain with enhancers


HIGHDEF® Hematoxylin is intended for use in the histologic demonstration of nuclear staining. This staining technique is sometimes used to make the critical distinction between a normal nucleus and an abnormal one.

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR