POLYVIEW® Plus HRP (Anti-Rabbit) Reagent, Enzo Life Sciences
Supplier: Enzo Life Sciences
ACC103-0150
75841-134EA 204.92 USD
75841-134
POLYVIEW® Plus HRP (Anti-Rabbit) Reagent, Enzo Life Sciences
Protein Linking and Labeling Systems
High sensitivity, low background nanopolymer detection reagent for use with HIGHDEF® chromogens in ISH and IHC applications.
  • Ready-to-use reagent
  • Biotin-free nanopolymer detection circumvents endogenous biotin background
  • High intensity color development delivers sharp, crisp staining
  • Compatible with both automated and manual platforms
  • The Antibody Blocker/Diluent further reduces background (sold separately)
  • Customize with HIGHDEF® chromogens of your choice (sold separately)
The POLYVIEW® Plus HRP (anti-rabbit) reagent is a ready-to-use, biotin-free, one-step detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, the POLYVIEW® Plus HRP (anti-rabbit) reagent can be adapted for in situ hybridization detection.