SAVIEW® Plus HRP Reagent, Enzo Life Sciences
Supplier: Enzo Life Sciences
ACC102-0150
75841-132EA 214.73 USD
75841-132
SAVIEW® Plus HRP Reagent, Enzo Life Sciences
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
High sensitivity, low background streptavidin-based nanopolymer detection reagent for use with HIGHDEF® chromogens in ISH and IHC applications.
  • Ready-to-use reagent
  • Streptavidin-based detection with reduced carbohydrate and biotin background
  • Suitable for both manual and automated platforms
  • Suitable for custom multiplex IHC protocols
  • The Streptavidin Blocker/Diluent further reduces background (sold separately)
  • Customize with HIGHDEF® chromogens of your choice (sold separately)
SAVIEW® Plus HRP reagent is a ready-to-use streptavidin-based nanopolymer detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, the SAVIEW® Plus HRP reagent can be used for in situ hybridization detection of biotinylated DNA/RNA probes.