Avantor OmniTop Assemblies, Pre-Sterilized Single-Use Sample Tubes®, Avantor Fluid Handling
Supplier: Avantor Fluid Handling
8017 8017 8017 8017 8017 8017 8017 8017 8017 8017
75840-754CS 4771.78 USD
75840-754 75840-756 75840-758 75840-760 75840-764 75840-766 75840-768 75840-770 75840-772 75840-774 75840-776
Avantor OmniTop Assemblies, Pre-Sterilized Single-Use Sample Tubes®, Avantor Fluid Handling
Tubes Test Tubes
OmniTop Sample Tubes® are a convenient device that can be used to obtain fluid samples. Each single-use OmniTop tube comes with a pre-attached 0.2 µm vent filter and 18" of tubing (C-Flex®, TYGON®, silicone or PharMed®).
  • Unique cap design allows for complete customization
  • Ability to use different Int.Ø and Ext.Ø tubing diameters
  • Reduced assembly and installation time
  • Eliminate cleaning validation
  • Flexible manufacturing
  • Design permits quick delivery on small lots
  • Closure system can be used with various glass bottles
These tubes are available with or without internal dip-tubes to facilitate removal of the fluid inside and has a wide selection of tubing materials. Easily configured with virtually any type of tubing.
OmniTop Sample Tubes® can be aseptically connected to bioreactor or other device using standard tubing welders, such as the Terumo SCD-IIB. Sample Tubes can also be aseptically sealed and removed (wet or dry) using tubing sealers such as the Genesis SE340 or handheld battery powered version SE640.
Use OmniTop for the transfer of biopharmaceutical products and reagents in a closed system, gas exchange transfer, sampling of biopharmaceutical products and reagents in a closed system, storage of biopharmaceutical products and reagents. For use with bench top, pilot plant, and production bioreactors. Also for use with sterile connecting devices and thermal or RF tubing sealers.
OmniTop Sample Tubes® are availble in a wide variety of standard configurations or they can be customized to suit specific applications.
Packaging: Available individually packaged or configured in manifold.