ActivArmr® 43-217 TIG Welding Gloves, Ansell
Supplier: Ansell Healthcare
43217100
75840-574CS 314.93 USD
75840-574 75840-576
ActivArmr® 43-217 TIG Welding Gloves, Ansell
Gloves
TIG welding glove for precise welding and thermal jobs.
  • Provides protection from heat, flames, sparks, and weld spatter
  • Premium goatskin for a relaxed, natural feel
  • Aramid stitching for improved durability and cost of ownership
  • Ideal for precise, technical welding applications
ActivArmr® 43-217 is a TIG welding glove that provides EN 12477 Type-B protection for precise welding and thermal tasks. It is suitable for TIG welding, as well as grinding, brazing, metal fabrication, and maintenance. These gloves provide protection from heat, flames, sparks, and weld spatter. The three-dimensional thumb design greatly increases fine dexterity for precise welding control. Premium full grain goatskin and welted seams with para-aramid thread ensure durability in use. The long-split cowhide cuff extends protection to the forearms.
ActivArmr® 43-217 provides comfort, dexterity, and protection in a cost effective thermal glove.