Leica EM TP Automatic Tissue Processor, Leica Microsystems
Supplier: Avantor
75840-174EA 18878.74 USD
75840-174 76414-912 76414-914
Leica EM TP Automatic Tissue Processor, Leica Microsystems
Tissue Processors
Automated routine tissue processor for EM and LM sample preparation.
  • Automated tissue processing
  • Closed processing chamber
  • High throughput
The innovative Leica EM TP is the first tissue processor designed for EM and LM resin processing that features a heating/cooling system with pre-heat and pre-cool capabilities to maintain a constant processing temperature.
It includes an easy-to-operate membrane control panel. The sample carousel holds 24 EM or 12 LM vials and utilizes various specialized baskets and capsules. An exhaust system supports safer use of toxic substances.
Caution: For research use only.