Midi Plus™ Pipetting Controller, Sartorius
Supplier: Sartorius
710931
75840-128EA 670.7 USD
75840-128
Midi Plus™ Pipetting Controller, Sartorius
Pipettes Pipette Controllers
The lightweight, cordless Midi Plus pipetting controller enables users to aspirate from bottles and tubes without holding their arms and hands in an uncomfortable raised position.
  • Precise finger tip control of variable speed ranges
  • Hydrophobic autoclavable filter prevents over aspiration
  • Low-battery warning
Users can achieve fine-tune speed by applying varying finger pressure to the operating buttons.
The pipetting controller fits all commonly used 1 to 100 ml glass or plastic pipettes.