Brady® Label Roll Adaptor for BMP®71 Label Printer, Brady Worldwide
Supplier: Brady Worldwide
147998
75840-084PK 32.71 USD
75840-084
Brady® Label Roll Adaptor for BMP®71 Label Printer, Brady Worldwide
Label and Tape Dispensers
Label Roll Adaptors enable the BMP®71 Label Printer to print on BMP®61, TLS 2200®, and HandiMark® Series printer labels and tapes.
  • Package contains three label roll adaptors (BMP®71 Label Printer and label rolls sold separately)
  • Expands the label offering for the BMP®71 Label Printer by more than 600 parts
  • Adaptor snaps on to BMP®61, TLS 2200®, and HandiMark® Series label rolls
This reusable spare adaptors save time by allowing users to keep them on the most frequently used label rolls